Learn More
Thermo Scientific™ Kit per la sintesi di cDNA DyNAmo™
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
:
100 Rxns
Descrizione
Thermo Scientific™ DyNAmo™ cDNA Synthesis Kit is optimized for quantitative reverse transcription PCR (qRT-PCR) applications, where amplicons are typically 100 bp in length.
The DyNAmo cDNA Synthesis Kit includes all necessary reagents for cDNA synthesis from various RNA sources. Total RNA, messenger RNA, viral RNA or in vitro transcribed RNA can all be used as a template for reverse transcription.
- Specifically optimized for quantitative RT-PCR
- Broad dynamic range: up to 1μg of RNA
- Reliable results with different RNA sources: mRNA, total RNA, viral RNA and in vitro transcribed RNA
- Flexible: applicable with all priming options for cDNA synthesis
- Easy to use: short protocol, 2x mix
Includes:
M-MuLV RNase H+ reverse transcriptase (includes RNase inhibitor), 2x RT buffer (includes dNTP mix and 10 mM MgCl2), Random hexamers (300 ng/μL), Oligo(dT)15 primer (100 ng/μL).
Recommended for:
cDNA synthesis for qRT-PCR
Specifica
Specifica
| Content And Storage | Trascrittasi inversa M-MuLV RNasi H+ (incluso inibitore di RNasi), tampone RT 2x (inclusa miscela dNTP e MgCl2 10 mM), esameri random (300 ng/µl), primer oligo(dT)15 (100 ng/µl). Conservare a -20 °C. |
| Product Type | Sintesi di cDNA per qRT-PCR |
| Quantità | 20 reazioni |
| Da utilizzare con (applicazione) | cDNA Synthesis |