Learn More
Thermo Scientific™ DNA polimerasi DyNAzyme I & II
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
1000 U (2 U/µl)
1000U (2U/µL)
250 U (2 U/µl)
Dimensione della confezione:
1000 unità
250 unità
Descrizione
DyNAzyme DNA Polymerases are Taq-like enzymes with improved thermal stability at high temperatures. They are ideal for all routine PCR applications, and have improved tolerance against DMSO. DyNAzyme I DNA Polymerase is a native thermostable DNA polymerase from Thermus brockianus. DyNAzyme II DNA Polymerase is an easy-to-use, recombinant thermostable DNA polymerase for standard PCR.
Highlights
- Improved thermal stability
- Improved tolerance against DMSO
- Supplied with optimized buffers or with Mg2+-free buffers and MgCl2 solution
- Detergent-free buffer also available
Specifica
Specifica
| Content And Storage | DyNAzyme II DNA Polymerase, 10x Optimized DyNAzyme BufferRecommended storage temperature -20 °C. Stable for one year from the assay date. |
| Descrizione | DyNAzyme™ II DNA Polymerase |
| Quantità | 1000U (2U/µL) |