Learn More
Trajan Scientific and Medical™ EC Science Silicon Septa
Designed for long lifetime and low injection port adhesion.
Marca: Trajan Scientific and Medical™ 041905
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Combines long injection lifetime, low bleed and low injection port adhesion
- Reliable seal against carrier gas pressure in the inlet
- No bleed or contamination into the GC system
- Maximum operating temperature of 350°C (300°C for 17mm size)
Specifica
EC | |
50/Pk. |