missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECHS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11305-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene functions in the second step of the mitochondrial fatty acid beta-oxidation pathway. It catalyzes the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs. The gene product is a member of the hydratase/isomerase superfamily. It localizes to the mitochondrial matrix. Transcript variants utilizing alternative transcription initiation sites have been described in the literature.Specifica
ECHS1 | |
Polyclonal | |
Unconjugated | |
ECHS1 | |
ECHS1, ECoH-1, Enoyl CoA hydratase 1, SCEH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140547, 1892, 93747 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P14604, P30084, Q8BH95 | |
ECHS1 | |
ECHS1 Fusion Protein Ag1842 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |