missing translation for 'onlineSavingsMsg'
Learn More

ECM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11521-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898653

  • 121.38€ / 20µL

Spedizione stimata: 03-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Secretory Component is an epithelial transport receptor for polymeric IgA and IgM. Antibody to secretory component has been shown to be useful for identifying glandular carcinomas.
Specifica

Specifica

ECM1
Polyclonal
Unconjugated
ECM1
ECM1, extracellular matrix protein 1, Secretory component p85
Rabbit
Antigen Affinity Chromatography
RUO
116662, 13601, 1893
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin)
0.19 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q16610, Q61508, Q62894
ECM1
ECM1 Fusion Protein Ag2084
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali