missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDG7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 19509-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The antibody is specific to LPAR3.
EDG7 is a member of the G protein-coupled receptor family, as well as the EDG family of proteins. This protein functions as a cellular receptor for lysophosphatidic acid and mediates lysophosphatidic acid-evoked calcium mobilization. This receptor couples predomitly to G(q/11) alpha proteins.EDG7 has been reported in human prostate, as well as rodent brain, kidney, lung, placenta, prostate, and testis. ESTs have been isolated from a normal prostate library, as well as cancer libraries of the head/neck, ovary, and thyroid.Specifica
EDG7 | |
Polyclonal | |
Unconjugated | |
LPAR3 | |
Edg 7, EDG7, GPCR, HOFNH30, LP A3, LPA 3, LPA receptor 3, LPA3, LPAR3, RP4 678I3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23566, 65086, 66025 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K5E0, Q9EQ31, Q9UBY5 | |
Lpar3 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |