Learn More
EGFR (Epidermal Growth Factor Receptor) Mouse, Clone: GFR/2341, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 1956-MSM11-P0
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| EGFR (Epidermal Growth Factor Receptor) | |
| Cell Surface | |
| GFR/2341 | |
| Unconjugated | |
| Mouse | |
| ∽170kDa (wild type) and ∽145kDa (vIII variant) | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 1956 | |
| IgG2b κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant fragment of human EGFR protein (apprx. aa 344-492) | |
| A431 cells. Placenta, Breast, Colon or Bladder cancer | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |