missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF2S3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11162-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The initiation of protein synthesis in eukaryotic cells is regulated by interactions between protein initiation factors and RNA molecules. The eukaryotic initiation complex is composed of three subunits, designated eIF2alpha, eIF2beta and eIF2gamma (eukaryotic translation initiation factor 2 alpha, beta and gamma, respectively), all of which work in concert to form a ternary complex with GTP and tRNA in the early stages of protein synthesis. eIF2S3 (eukaryotic translation initiation factor 2, subunit 3), also known as eIF2G, is a 472 amino acid protein that belongs to the gamma subfamily of GTP-binding elongation factor proteins. Existing as a heterotrimer composed of an alpha, beta and gamma chain, eIF2S3 functions to bind ribosomal subunits and catalyze the subsequent formation of preinitiationcomplexes necessary for protein synthesis.Specifica
EIF2S3 | |
Polyclonal | |
Unconjugated | |
EIF2S3 | |
eIF 2 gamma X, eIF 2gA, eIF 2gX, EIF2, EIF2G, EIF2gamma, EIF2S3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1968 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P41091 | |
EIF2S3 | |
EIF2S3 Fusion Protein Ag1650 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |