missing translation for 'onlineSavingsMsg'
Learn More

EIF5A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 17069-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16896894

  • 121.38€ / 20µL

Spedizione stimata: 03-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

mRNA-binding protein involved in translation elongation. Has an important function at the level of mRNA turnover, probably acting downstream of decapping. Involved in actin dynamics and cell cycle progression, mRNA decay and probably in a pathway involved in stress response and maintece of cell wall integrity. Functions as a regulator of apoptosis. Mediates effects of polyamines on neuronal process extension and survival. May play an important role in brain development and function, and in skeletal muscle stem cell differentiation.
Specifica

Specifica

EIF5A2
Polyclonal
Unconjugated
EIF5A2
eIF 5A 2, EIF 5A2, EIF5A2, eIF5AII
Rabbit
Antigen Affinity Chromatography
RUO
208691, 310261, 56648
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BGY2, Q9GZV4
Eif5a2
EIF5A2 Fusion Protein Ag10895
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali