Learn More
Merck MultiScreen™HTS 96-Well Filter Plates for Clearing
Designed to meet specific performance criteria for sample preparation methodology
Marca: Merck MSNANLY10
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Automation-compatible multiscreen filter plates
- Lysate clearing
Clearing of bacterial cell lysates
Specifica
Standard | |
Clear | |
96 | |
Clearing of bacterial cell lysates |
8 x 12 | |
Styrene | |
Non-sterile | |
10 |