Learn More
Merck Pellicon™ XL Devices
Small-volume tangential flow filter device
Marca: Merck PXB500C50
Dettagli aggiuntivi : Peso : 0.50000kg
Includes: Operating instructions and Certificate of Quality
Descrizione
Designed for process development and sample preparation, with an optimal processing volume range of 50 to 1000mL.
- True linear scalability with same flow path, channel length, and channel height as other established Pellicon products
- Can operate with peristaltic pump or Labscale™ TFF system Robust thermoplastic (polypropylene) construction
- Controlled channel height
- Choice of Biomax™ PES (polyethersulfone) or Ultracel™ PLC (composite regenerated cellulose) membranes
Concentration and desalting of proteins, buffer exchange, depyrogenation, clarification, and preparation of material for clinical trials.
Order Info
Packaged one per box.
Specifica
Ultrafiltration Biomax-500 | |
Polypropylene | |
100 μm | |
Protein purification, analytical sample preparation | |
80psi at 50°C | |
1 Pack | |
Pellicon XL Ultrafiltration Device | |
3.2 mL | |
7.4 x 1.2 in. (18.8 x 3cm) | |
18.8 cm | |
3 cm |
Cassette | |
500 kDa | |
50 cm2 | |
Operating instructions and certificate of quality | |
1 to 14 | |
0.03 to 0.05L/min. | |
0.5 oz. | |
15 mL | |
7.4 in. | |
1.2 in. |