Learn More
Memmert™ Entry Port Left Centre/Centre
For introducing connections at the side. Moisture tight and can be closed by flap and silicone stopper
Marca: Memmert™ HPP410ecoF0
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- Standard position left centre/centre
- Not for model size 260 with light module
- Not for model size 110 with light module
Specifica
Moisture tight | |
HPP410eco |