Learn More
Memmert™ Entry Port Right (Individual Location)
Entry port with flap closure and custom TBC position in right side
Marca: Memmert™ HPP1400ecoF5
Dettagli aggiuntivi : Peso : 0.01100kg
Descrizione
- Not in combination with illumination units
- Moisture tight
- Can be closed by flap and silicone stopper
Specifica
Clear | |
HPP1400eco | |
23 mm |