Learn More
Cytiva (formerly Pall Lab) Envirochek™ Supor™ Sampling Capsule
Envirochek™ capsules are approved for concentration and recovery of Cryptosporidium oocysts and Giardia cysts in water.
Brand: Cytiva (formerly Pall Lab) 12107
Additional Details : Peso : 4.26550kg
Description
- Approved for concentration and recovery of Cryptosporidium oocysts and Giardia cysts in water according to EPA method 1623.1, with a faster, safer, and simpler protocol than other methods including processing of up to 8 samples at once using our shaker
Specifications
Urethane Adhesive | |
1 μm | |
1300 cm2 | |
25/Pk., Individually Pagged |
Hydrophilic PES | |
Non-sterile | |
Polycarbonate | |
Straight Hose Barb |