Learn More
Epcam / CD326 Mouse, Clone: EGP40/1384, enQuireBio™
Mouse Monoclonal Antibody
Marca:  EnquireBio 4072-MSM14-P1
Dettagli aggiuntivi : Peso : 0.01000kg
Codice prodotto. 15950418
  • 462.00€ / 100µg
Spedizione stimata: 03-02-2023
per visualizzare lo stock disponibile
Specifica
Epcam / CD326
Cell surface
EGP40/1384
Unconjugated
NP_002345
Mouse
40-43kDa
Protein A/G Purified
RUO
4072
IgG
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
EPCAM
Recombinant human EpCAM protein fragment from the extracellular domain (aa77-202)
HT29 cells or Ovarian carcinoma
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.