Learn More
Brady™ ES Type 10 Tag Out
Marca: Brady™ 284429
Dettagli aggiuntivi : Peso : 0.06500kg
Specifica
ES Type 10 Tag Out | |
Polyester | |
5.71 in. | |
10 Pk. | |
-40°C to +100°C | |
3.35 in. |
White on Green | |
Safety Identification | |
145 mm | |
-40°F to +212°F | |
0.249 mm | |
85 mm |