Learn More
Ethyl 3-methylvalerate, 98%, Thermo Scientific™
Catalog Number 11493118
Click to view available options
Quantità:
5 g
25 g
5870-68-8
C8H16O2
144.214
MFCD00041390
TXAWGHYFBQBVNK-UHFFFAOYSA-N
ethyl 3-methylvalerate, pentanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylvalerate, valeric acid, 3-methyl-, ethyl ester, 3-methylvaleric acid ethyl ester, 3-methylpentanoic acid, ethyl ester, fema no. 3679, ethyl 3-methyl valerate, ethyl 3-methyl pentanoate
560255
ethyl 3-methylpentanoate
CCC(C)CC(=O)OCC
5870-68-8
C8H16O2
144.214
MFCD00041390
TXAWGHYFBQBVNK-UHFFFAOYSA-N
ethyl 3-methylvalerate, pentanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylvalerate, valeric acid, 3-methyl-, ethyl ester, 3-methylvaleric acid ethyl ester, 3-methylpentanoic acid, ethyl ester, fema no. 3679, ethyl 3-methyl valerate, ethyl 3-methyl pentanoate
560255
ethyl 3-methylpentanoate
CCC(C)CC(=O)OCC
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Ethyl 3-methylvalerate
Densità 0.87
Punti di ebollizione 158°C
Punto d'infiammabilità 45°C (113°F)
Odore Ester-like
Indice di rifrazione 1.407
Quantità 5 g
Numero UN UN3272
Peso formulazione 144.21
Percent Purity 98%