Learn More
Memmert™ Extended Overtemperature Protection for SF110
Extended overtemperature protection with integrated Pt100 sensor for independent temperature monitoring.
Marca: Memmert™ SF110A6
Dettagli aggiuntivi : Peso : 0.01200kg
Descrizione
- Accessory
- With integrated Pt100 sensor
- For models with SingleDISPLAY
Specifica
SF110 Sterilizer | |
Integrated Pt100 Sensor |