Learn More
Lenz Laborglasintrumente Extractors Acc. to Soxhlet with Allihn Condenser
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca: Lenz Laborglasintrumente 05365043
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Extractor in compact and robust design
- With flask connection
- Made of DURAN™ tubing
- Allihn condenser with thread GL 14
- Round bottom flask
Specifica
150 mL | |
Duran Glass | |
NS 45/40 |