Learn More
FAM49B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20127-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
| FAM49B | |
| Polyclonal | |
| Unconjugated | |
| FAM49B | |
| BM 009, FAM49B, L1, Protein FAM49B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 223601, 299909, 51571 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q921M7, Q9NUQ9 | |
| FAM49B | |
| FAM49B Fusion Protein Ag13939 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |