Learn More
FANCD2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 28619-1-AP-20UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
The FANCD2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FANCD2. This antibody recognizes human antigen. The FANCD2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifica
| FANCD2 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 2177 | |
| IgG |
| Western Blot, ELISA | |
| 400 μg/mL | |
| Rabbit | |
| Primary | |
| Human |