Learn More
Scotch-Brite™ Fast Shine Microfiber Wipes 2060
Semi-disposable microfiber wipes with great cleaning properties. Offers quick and effective cleaning performance when used with or without chemicals on a range of surfaces.
Marca: Scotch-Brite™ 7000033497
Dettagli aggiuntivi : Peso : 0.01800kg
Descrizione
- Quick and effective cleaning performance when used with or without chemicals
- Lint free cleaning on virtually any surface
- Eliminates the need for cleaning agents in many cases
- Ideal for cleaning furniture and sanitary ware
- Can be used on all types of surfaces, including fragile surfaces
Specifica
Wipes | |
360 mm | |
Green | |
400 x 360 mm | |
10/Pk. |
400 mm | |
Microfiber | |
Semi-disposable | |
Quick and Effective Cleaning |