missing translation for 'onlineSavingsMsg'
Learn More

FBXW5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 18321-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16898394

  • 140.00€ / 20µL
Spedizione stimata: 15-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a member of the F-box protein family, members of which are characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into three classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene contains WD-40 domains, in addition to an F-box motif, so it belongs to the Fbw class. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene, however, they were found to be nonsense-mediated mRNA decay (NMD) candidates, hence not represented.
Specifica

Specifica

FBXW5
Polyclonal
Unconjugated
FBXW5
Fbw5, FBXW5, RP11 229P13.10
Rabbit
Antigen Affinity Chromatography
RUO
30839, 362081, 54461
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q4KLI9, Q969U6, Q9QXW2
Fbxw5
FBXW5 Fusion Protein Ag13163
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali