Learn More
FCHSD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA560514
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Immunogen sequence: AHLDHYYQEE LPALLKALVS ELSEHLRDPL TSLSHTELEA AEVILEHAHR GEQTTSQVSW EQDLKLFLQE PGVFSPTPPQ QFQPAGTDQV CVLEWG Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 81%.
FCHSD1 gene ontology annotations related to this gene include neuromuscular synaptic transmission; regulation of actin filament polymerization.Specifica
FCHSD1 | |
Polyclonal | |
Unconjugated | |
FCHSD1 | |
A030002D08Rik; F-BAR and double SH3 domains protein 1; FCH and double SH3 domains 1; FCH and double SH3 domains protein 1; Fchsd1; FLJ00007; nervous wreck homolog 2; NWK2; Protein nervous wreck 2; UNQ737/PRO1431 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
89848 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86WN1 | |
FCHSD1 | |
Recombinant protein corresponding to Human FCHSD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only