missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FCRN/FCGRT Antibody (nipocalimab), Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals™ NBP3-28599-50ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
FCRN/FCGRT Monoclonal antibody specifically detects FCRN/FCGRT in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifica
| FCRN/FCGRT | |
| Monoclonal | |
| Unconjugated | |
| P55899 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| nipocalimab | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig | |
| FcRn (FCGRT & B2M) | |
| 50 μg | |
| Immunology | |
| 2217 | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto