missing translation for 'onlineSavingsMsg'
Learn More

FGF12 Mouse anti-Human, Clone: 2F11A10, Proteintech

Mouse Monoclonal Antibody

Marca:  Proteintech 60152-1-IG-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16897115

  • 349.00€ / 150µL

Spedizione stimata: 16-12-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth, and invasion. This growth factor lacks the N-terminal signal sequence present in most of the FGF family members, but it contains clusters of basic residues that have been demonstrated to act as a nuclear localization signal. When transfected into mammalian cells, this protein accumulated in the nucleus, but was not secreted. The specific function of this gene has not yet been determined. Two alternatively spliced transcript variants encoding distinct isoforms have been reported.
Specifica

Specifica

FGF12
Monoclonal
1.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P61328
FGF12
FGF12 Fusion Protein Ag5046
150 μL
Primary
Human
Antibody
IgG2b
Western Blot
2F11A10
Unconjugated
FGF12
FGF 12, FGF12, FGF12B, FHF 1, FHF1, fibroblast growth factor 12, Myocyte activating factor
Mouse
Caprylic Acid Precipitation
RUO
2257
-20°C
Liquid
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali