missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11375-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descripción
This gene encodes a protein that is localized in the mitochondrial inner compartment and that may play a role in mitochondrial apoptosis. Nonsense mutations have been reported to result in cytochrome c oxidase deficiency.Especificaciones
FH | |
Polyclonal | |
Unconjugated | |
FH | |
FH, Fumarase, fumarate hydratase, HLRCC, LRCC, MCL, MCUL1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14193, 2271, 24368 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P07954, P14408, P97807 | |
FH | |
FH Fusion Protein Ag1934 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |