missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Filaggrin (Keratinocyte Differentiation Marker) Mouse, Clone: FLG/1563, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 2312-MSM3-P1
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Filaggrin (Keratinocyte Differentiation Marker) | |
Cytoplasmic | |
FLG/1563 | |
Unconjugated | |
Mouse | |
26-45kDa (Processed); 350kDa (Profilaggrin) | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
2312 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human Filaggrin protein fragment (apprx. aa 998-1104) | |
U-251-MG cells or Skin | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |