Learn More
ROBU VitraPOR™ Filter Funnel
Filter funnel available in a buchner shape or a conical shape.
Marca: ROBU VitraPOR™ 21250
Dettagli aggiuntivi : Peso : 0.27840kg
Descrizione
- Highly resistant to most chemicals and temperatures up to 500°C
Specifica
250 mL | |
Borosilicate Glass 3.3 | |
Buchner Shape | |
0 |