Learn More
Greiner Bio-One Filter Tips for Gilson™ Pipetman Pipettes
Prevents contamination with liquids and aerosols during pipetting process.
Marca: Greiner Bio-One 771288
Dettagli aggiuntivi : Peso : 1.33000kg
Descrizione
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Non-cytotoxic
- Prevents aspiration of particles into interior of pipette
- Minimizes danger of unwanted cross-contamination
- Works with DNA and RNA and ra
Specifica
10 μL | |
Sterile | |
Irradiation | |
Gilson™ Pipetman™ P2, Gilson™ Pipetman™ P10 | |
Polypropylene |
Filtered | |
Translucent | |
Micro Point | |
No |