missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FIP1L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12276-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
FIP1L1 is a component of the cleavage and polyadenylation specificity factor (CPSF) complex that plays a key role in pre-mRNA 3′-end formation, recognizing the AAUAAA signal sequence and interacting with poly(A) polymerase and other factors to bring about cleavage and poly(A) addition. FIP1L1 contributes to poly(A) site recognition and stimulates poly(A) addition. It binds to U-rich RNA sequence elements surrounding the poly(A) site. FIP1L1 may act to tether poly(A) polymerase to the CPSF complex.Especificaciones
| FIP1L1 | |
| Polyclonal | |
| Unconjugated | |
| FIP1L1 | |
| Factor interacting with PAP, FIP1, FIP1 like 1 (S. cerevisiae), FIP1 like 1 protein, FIP1L1, FIP1L1 PDGFRalpha, hFip1, Rhe | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 289582, 66899, 81608 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5U317, Q6UN15, Q9D824 | |
| Fip1l1 | |
| FIP1L1 Fusion Protein Ag2921 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido