Learn More
Fisherbrand™ Vortex Mixer Accessories: Foam Racks
For use with Fisherbrand vortex mixers.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
- Mixes 96-well plates or deepwell blocks
- Requires Insert Retainer (02-215-391)
Specifica
Microplate Holder | |
Foam | |
Digital and analog mini vortex mixer |