missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems FITC-Labeled Human CD27 Ligand / CD70 Protein, His,Flag Tag active trimer

MALS verified

Marca:  ACROBiosystems CDD-HF2D4-200ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17106221

  • 2992.91€ / 200µg
Spedizione stimata: 11-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Cluster of Differentiation 70 (CD70) is also known as CD27 ligand (CD27L / CD27LG), TNFSF7, TNFSF7G, is a type II transmembrane glycoprotein belonging to the TNF superfamily (TNFSF) and is a surface antigen found on activated T-and B-lymphocytes and mature dendritic cells. Binding of CD70 to its receptor CD27 induces in priming, effector functions, differentiation and memory formation of T-cells, and thus is invloved in the biological processes including T-cell activation, the proliferation of costimulated T-cells, as well as the generation of cytolytic T-cells. CD70 on T cells provides costimulatory signals that are required for T cell proliferation, clonal expansion and the promotion of effector T cell formation. CD70 on mouse B cell has been shown to inhibit terminal differentiation of activated B cells into plasma cells and enhances commitment to memory B cell responses. CD70 induces proliferation and IFNγ production, on NK cells.

This protein carries a polyhistidine tag at the N-terminus, followed by a flag tag. The protein has a calculated MW of 49.3 kDa. The protein migrates as 50-55 kDa under non-reducing (NR) condition (SDS-PAGE) due to glycosylation.
Specifica

Specifica

49.3 kDa
FITC-Labeled Human CD27 Ligand, His,Flag Tag (CDD-HF2D4) is expressed from human 293 cells (HEK293). It contains AA Ser 52 - Pro 193 (Accession # P32970-1). It is the biotinylated form of Human CD27 Ligand, His,Flag Tag (active trimer) (MALS verified) (Cat. No.CDL-H52Da).
-20°C, avoid light
CD27 Ligand
Human
PBS, pH7.4
HEK293
Fluorescent labeled protein
200 μg
Ser 52 - Pro 193
CD70,CD27LG,TNFSF7,TNFSF7G,CD27L
FITC
His Tag and Flag Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali