Learn More
Dostmann Electronic™ Flash III Infrared Thermometer
Dostmann Electronic™ Flash III Infrared Thermometer is a two-in-one thermometer featuring infrared and probe measuring options.
Marca: Dostmann Electronic™ 5020-0556
Dettagli aggiuntivi : Peso : 0.11000kg
Descrizione
- Temperature measurement via contact (probe) or non-contact (infrared) methods
- Display of current, highest or lowest temperature
- HACCP-LED indicates HI/LO-Level for food industry measurements
- Waterproof IP 65
- MAX-MIN-HOLD-LOCK-function
- °C/°F-switchable
- LED spot size indicator
- Clip for fixing, bag, and calibration certificate included
Specifica
0.01 to 1.00 | |
Flash 3 Infrared Thermometer | |
-33°C (Infrared), -55°C (Insertion Probe) | |
Infrared: -33°C to 0°C ±1.0°C to ±0.1/1°C 0°C to +65°C + |
0.1°C | |
220°C | |
-33°C to +220°C (Infrared), -55°C to +220°C (Insertion Probe) |