missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLI1 Antibody (FLI1/4371R), Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP3-20544-20ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
FLI1 Monoclonal antibody specifically detects FLI1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifica
| FLI1 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| EWSR2, Friend leukemia integration 1 transcription factor, Friend leukemia virus integration 1, Proto-oncogene Fli-1, SIC-1, Transcription factor ERGB | |
| Recombinant full-length human FLI1 protein | |
| 20 μg | |
| Cancer, Tumor Suppressors | |
| 2313 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| FLI1/4371R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction