Learn More
Jenway™ Fluorimeter Interference Filter
Requires a much more narrow range of wavelengths. Jenway™ Fluorimeter Interference Filter allows a specified wavelength and light to pass through to the sample.
Marca: Jenway™ 10089-77
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- For use with Jenway™ Fluorimeters
- Normally used as the emission filters where a much narrower range of wavelengths is required
- Allow the specified wavelength and light ±5nm of this through to the sample.
Specifica
Interference Filter | |
370 nm | |
Jenway Fluorimeters |