Learn More
Lenz Laborglasinstrumente™ Palloni a quattro colli
Especially made for reactions and can be used to make solutions or hold, contain and collect chemicals. Lenz Laborglasinstrumente™ Four-Neck Round Bottom Flasks, Side Necks Parallel are manufactured from durable DURAN™ borosilicate glass.
Marca: Lenz Laborglasinstrumente™ B0425991
Dettagli aggiuntivi : Peso : 0.30600kg
Descrizione
- Acc. to DIN 12392
- With three parallel necks at a 90° angle
- With inscription label
- Corrosion-free and resistant to thermal shock
- With conical joints, featuring high taper accuracy and excellent tightness
Specifica
500 mL | |
1 ciascuno | |
Rotondo |
A quattro colli | |
NS 45 |