missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FYN Mouse anti-Human, Mouse, Rat, Clone: 1G4B1, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66606-1-IG-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descripción
This gene is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist.Especificaciones
| FYN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P06241, P39688, Q62844 | |
| Fyn | |
| FYN Fusion Protein Ag11127 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1G4B1 | |
| Unconjugated | |
| FYN | |
| FYN, p59 Fyn, Proto oncogene c Fyn, Proto oncogene Syn, SLK, Src like kinase, SYN, Tyrosine protein kinase Fyn | |
| Mouse | |
| Protein G | |
| RUO | |
| 14360, 25150, 2534 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido