missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GALT Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17035-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Galactose-1-phosphate uridyl transferase (GALT) catalyzes the second step of the Leloir pathway of galactose metabolism, namely the conversion of UDP-glucose + galactose-1-phosphate to glucose-1-phosphate + UDP-galactose. The absence of this enzyme results in classic galactosemia in humans and can be fatal in the newborn period if lactose is not removed from the diet. The pathophysiology of galactosemia has not been clearly defined.Specifica
GALT | |
Polyclonal | |
Unconjugated | |
GALT | |
Gal 1 P uridylyltransferase, GALT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2592 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P07902 | |
GALT | |
GALT Fusion Protein Ag10524 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |