missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gastrokine 1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Specifica
Antigene | Gastrokine 1 |
---|---|
Concentrazione | 0.43 mg/mL |
Applicazioni | Western Blot, Immunohistochemistry (Paraffin) |
Classificazione | Polyclonal |
Coniugato | Unconjugated |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
16898954
|
Proteintech
19344-1-AP-20UL |
20 μL |
140.00€
20µL |
Spedizione stimata: 14-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
16888954
|
Proteintech
19344-1-AP-150UL |
150 μL |
399.00€
150µL |
Spedizione stimata: 14-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
Gastrokine 1, also known as GKN1 and CA11, is a 185 amino acid containing secreted protein belonging to the gastrokine family with a BRICHOS domain and an asn-rich region. GKN1 (also known as an antioncogene) is reported to be down-regulated in human gastric cancer tissue as compared to normal gastric mucosa. Differential display of normal and cancerous gastric tissue suggested that this protein is predomitly expressed in stomach and the expression is down-regulated to considerable frequency in cancer tissue or gastric cancer cell lines examined. This extra cellular protein is co-secreted with mucins and is a signaling growth factor with mitogenic activity and may be involved in maintaining the integrity of the gastric mucosal epithelium.Specifica
Gastrokine 1 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q9CR36, Q9NS71 | |
56287, 66283 | |
Gastrokine 1 Fusion Protein Ag6091 | |
Primary | |
-20°C | |
GKN1 |
0.43 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
18 kDa antrum mucosa protein, AMP 18, AMP18, BRICD1, CA11, FOV, foveolin, gastrokine 1, GKN1, Protein CA11 | |
GKN1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |