missing translation for 'onlineSavingsMsg'
Learn More

GFM1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA557050

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15724951

  • 477.00€ / 100µL
Spedizione stimata: 16-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: QYGKVIGVLE PLDPEDYTKL EFSDETFGSN IPKQFVPAVE KGFLDACEKG PLSGHKLSGL RFVLQDGAHH MVDSNEISFI RAGE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. Mitochondrial translation is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintenance of mitochondrial DNA. This gene encodes one of the mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known.
Specifica

Specifica

GFM1
Polyclonal
Unconjugated
GFM1
AW545374; COXPD1; D3Wsu133e; Efg; EF-G; Efg1; EFGM; EF-Gmt; EF-Gmt {ECO:0000255; EGF1; Elongation factor G 1, mitochondrial; elongation factor G 1, mitochondrial {ECO:0000255; elongation factor G, mitochondrial; elongation factor G, mitochondrial {ECO:0000255; Elongation factor G1; elongation factor G1 {ECO:0000255; G elongation factor 1; G elongation factor mitochondrial 1; G elongation factor, mitochondrial 1; G translation elongation factor, mitochondrial; GFM; Gfm1; HAMAP-Rule:MF_03061}; hEFG1; mEF-G 1; mEF-G 1 {ECO:0000255; mitochondrial; mitochondrial elongation factor G; mitochondrial elongation factor G1
Rabbit
Antigen affinity chromatography
RUO
85476
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry, Western Blot
Lot-Specific
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96RP9
GFM1
Recombinant protein corresponding to Human GFM1
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only