missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NB600-308-1mg
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
GFP Polyclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Dot Blot, Electron Microscopy, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockdown Validated.
Specifica
| GFP | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| anti-gfp (green fluorescent protein) pab, Enhanced Green Fluorescent Protein, GFP immunofluorescence, GFP immunoprecipitation, GFP mouse, GFP staining, GFP western blot, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| Rabbit | |
| 27 kDa | |
| 1 mg | |
| Cellular Markers, Epitope Tags, Fusion Proteins, Mitochondrial Fusion Proteins, Mitochondrial Markers | |
| No reaction was observed against Human, Mouse or Rat serum proteins. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Immunohistochemistry (Free Floating) | |
| 0.99 mg/mL | |
| Western Blot 1:500-1:5000, Flow Cytometry 1:10-1:1000, ELISA 1:20000-1:120000, Immunohistochemistry 1:200-1:3000, Immunocytochemistry/Immunofluorescence 1:500-1:5000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:250, Dot Blot, Electron Microscopy 1:10-1:500, Immunohistochemistry Free-Floating, Immunohistochemistry, KnockDown Validated | |
| P42212 | |
| GFP | |
| The immunogen is a Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246aa) derived from the jellyfish Aequorea victoria. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Non-species specific, C. elegans, Plant | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto