Learn More
Biochrom Glass Cuvette, Flow Through Cell
Glass Cuvette, Flow Through Cell is a 10 mm Optical Glass cell with 450 μL working volume and 10 mm pathlength.
Marca: Biochrom 80-2004-45
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Optical Glass construction
- Flow Through Cell
- Working Volume: 450 μL
- 10mm pathlength
- Fits standard spectrophotometer cell holders
Specifica
Flow-Through Cell | |
Glass | |
1 Ea. | |
Ultrospec 7000/8000/9000 and Ultrospec 7500 and 2100 Pro |
450 μL | |
10 mm | |
10 mm |