Learn More
Lenz Laborglasintrumente Glass Filter Plugs with Tube
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca: Lenz Laborglasintrumente 07208420
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Filter plugs
- Cylindrical
- With tube (dia. 8 mm x length 210 mm)
- Material: borosilicate glass 3.3
Specifica
Borosilicate Glass (3.3) | |
4 | |
210 mm | |
9 mm |