Learn More
Hirschmann™ Fiasca volumetrica in vetro senza stopper
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with NS Ground are manufactured from durable DURAN™ glass tubing.
Marca: Hirschmann™ 2820370
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
- With NS ground, blue graduation
- Without stopper
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch information
Specifica
DIN EN ISO 1042 | |
Vetro Duran, Duran Glass | |
±0.04 ml | |
±0.04 ml | |
A |
25 ml | |
110 mm | |
Smeriglio standard 10/19 NS | |
Sì |