missing translation for 'onlineSavingsMsg'
Learn More

Labconco™ Glassware Washer Base Stand

Accessories for use with Labconco Glassware Washers with CleanWorks OS.

Marca:  Labconco™ 4653300

Dettagli aggiuntivi : Peso : 47.62720kg

 vedi altre versioni di questo prodotto

Codice prodotto. 17102352

  • 1865.00€ / Pezzo

Spedizione stimata: 22-02-2023
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

  • Racks and accessories for use with Labconco FlaskScrubber, SteamScrubber, FlaskScrubber Vantage Glassware Washers
  • Base Stands elevate freestanding SteamScrubber and FlaskScrubber or FlaskScrubber Vantage washers 17 in. for loading convenience
  • Includes one storage compartment with hinged door
  • Type 304 stainless steel
  • Stationary stand, fully assembled with leveling feet
Specifica

Specifica

FlaskScrubber Vantage Washer Base Stand
Washer Base Stand
Labconco FlaskScrubber Vantage
63.8 cm
43.2 cm
63.8 cm
81.3 cm
47.6 kg
Stainless Steel
1 washer
25.1 in.
17 in.
25.1 in.
32 in.
105 lb.
1/Ea.
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali