missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione S-Transferase/GST Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB9499-SP
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Glutathione S-Transferase/GST Monoclonal specifically detects Glutathione S-Transferase/GST in Multi-Species samples. It is validated for Western Blot.
Specifica
| Glutathione S-Transferase/GST | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Detects Glutathione S-Transferase/GST in direct ELISAs. | |
| Multi-species | |
| Purified |
| Western Blot | |
| 965427 | |
| Western Blot 0.25 ug/mL | |
| P08515 | |
| E. coli-derived recombinant Glutathione S-Transferase/GST epitope tag Accession # P08515 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
RUO
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto