missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMNN Mouse anti-Human, Clone: 2E3A2, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66566-1-IG-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a protein that plays a critical role in cell cycle regulation. The encoded protein inhibits DNA replication by binding to DNA replication factor Cdt1, preventing the incorporation of minichromosome maintece proteins into the pre-replication complex. The encoded protein is expressed during the S and G2 phases of the cell cycle and is degraded by the anaphase-promoting complex during the metaphase-anaphase transition. Increased expression of this gene may play a role in several maligcies including colon, rectal and breast cancer. Alternatively spliced transcript variants have been observed for this gene, and two pseudogenes of this gene are located on the short arm of chromosome 16.Specifica
GMNN | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75496 | |
GMNN | |
GMNN Fusion Protein Ag24283 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
2E3A2 | |
Unconjugated | |
GMNN | |
Gem, Geminin, GMNN, RP3 369A17.3 | |
Mouse | |
Protein G | |
RUO | |
51053 | |
-20°C | |
Liquid |