Learn More
GMPPA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15517-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
GMPPA is a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides. This gene is thought to encode a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides.Specifica
| GMPPA | |
| Polyclonal | |
| Unconjugated | |
| GMPPA | |
| GMPPA | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29926, 501167, 69080 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5XIC1, Q922H4, Q96IJ6 | |
| Gmppa | |
| GMPPA Fusion Protein Ag7789 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |