missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMPR 1/2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15683-1-AP-150UL
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
This antibody can recognize both GMPR1 and GMPR2 due to the high homology.
This gene encodes an enzyme that catalyzes the irreversible and NADPH-dependent reductive deamination of GMP to IMP. The protein also functions in the re-utilization of free intracellular bases and purine nucleosides.Specifica
| GMPR 1/2 | |
| Polyclonal | |
| Unconjugated | |
| GMPR | |
| GMP reductase 1, GMPR, GMPR1, GMPR2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 105446, 117533, 192357, 2766, 51292, 66355 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P36959, Q99L27, Q9DCZ1, Q9P2T1, Q9Z244 | |
| GMPR, GMPR2 | |
| GMPR 1/2 Fusion Protein Ag8185 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Spot an opportunity for improvement?Share a Content Correction